Bottles, wide neck, with screw cap, VWR®
Supplier: VWR, part of Avantor
VWR®
215-5636EA 43.7 EUR
215-5636 215-5635 215-5634 215-5633 215-5632 215-5631 215-5630
Bottles, wide neck, with screw cap, VWR®
Bottles Plastic Bottles
LDPE, translucent.
  • Good chemical resistance
  • Leakproof, tough and versatile
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The wide mouth allows easy filling and cleaning. Without graduations.