Bottles, wide neck, with screw cap, VWR®
Supplier: VWR, part of Avantor
VWR®
215-5679EA 39.8 EUR
215-5679 215-5678 215-5677 215-5676 215-5680
Bottles, wide neck, with screw cap, VWR®
Bottles Plastic Bottles
LDPE.
  • Amber coloured to protect light-sensitive substances
  • Leakproof, tough and versatile
Sample collection, storage and transport, for liquids, pastes and granular material. The wide mouth allows easy filling and cleaning.