Spray bottles, LaboPlast®
Supplier: Bürkle
LaboPlast®
215-6510EA 5.58 EUR
215-6510 215-6511 215-6513
Spray bottles, LaboPlast®
Bottles Spray Bottles
LDPE, translucent, PP spray attachment.
  • Good chemical resistance, good temperature resistance
  • Spray nozzle adjustment can be varied by rotation
  • Pressureless spray bottle
  • Media-contacting parts PP, PE and stainless steel
Spray, choice of fine mist or focused jet spray functions to disperse fluids.