Bottles, wide neck, square, with screw cap, VWR®
Supplier: VWR, part of Avantor
215-7396EA 7.1 EUR
215-7396 215-7390 215-7394 215-7392
Bottles, wide neck, square, with screw cap, VWR®
Bottles Plastic Bottles
HDPE with black, HDPE, tamper-evident cap.
  • Good chemical resistance
  • Leakproof
  • Space saving
Perfect for sample collection, storage and transport. For liquids, pastes and granular material. The wide mouth allows easy filling and cleaning.