VWR®, Dishes, Evaporating, Shallow
Supplier: VWR
VWR®
216-0173EA 28.8 EUR
216-0173 216-0176 216-0177 216-0174 216-0175 216-0178
VWR®, Dishes, Evaporating, Shallow
Trays/Dishes/Pots Evaporating Basins/Dishes
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Thick walled, conical shape. Extremely robust.