Dropping bottles, PTFE, VWR®
Supplier: VWR, part of Avantor
VWR®
216-0457EA 51 EUR
216-0457 216-0456
Dropping bottles, PTFE, VWR®
Bottles Dropping Bottles
PTFE, white.
  • Flexible wall for easy use
  • Screw-fit cap
  • Guaranteed chemically resistant and inert
Suitable for dropping-out aggressive chemicals.