Carboys, wide neck, HDPE
Supplier: VWR, part of Avantor
VWR®
216-0635EA 219 EUR
216-0635 216-0628 216-0621 216-0622 216-0642 216-0615 216-0629 216-0634
Carboys, wide neck, HDPE
Bottles Carboys
HDPE.
  • Versatile Cap technology provides the flexibility of interchangeable connectors
  • Extra wide mouth version for easy filling and cleaning
  • Smart Barb technology provides interchangeable spigot options
The VWR HDPE carboy is an advanced fluid handling system developed with the end user in mind. The ergonomically-designed carboy body, coupled with our VWR® Versatile Cap technology, provides the user with something that is easy to use and easy to clean. Made from HDPE makes it more scratch resistant and better resistant to chemicals than other carboys, making them ideal for most buffers and reagents, as well as powders and solids. The carboy body includes design features that allow the user to easily hold and manipulate during lifting, filling, and pouring, while VWR® Versatile Cap technology provides interchangeable, leakproof top connections. Features include a set of handles at both the top and bottom, easy to read embossed graduation marks, and moulded carboy material type identification mark. 20 and 40 litre carboys are also available with an extra wide mouth for easier filling and cleaning.