Carboys, wide neck, rectangular, with stopcock, VWR®

Supplier: VWR, part of Avantor
VWR®
216-1732EA 112 EUR
216-1732 216-1700 216-1719 216-1735
Carboys, wide neck, rectangular, with stopcock, VWR®
Bottles Carboys
HDPE or PP translucent, with purple screw caps.

Use for storing and dispensing solutions and media. Rectangular carboys feature a heavy-duty stainless steel handle for easy carrying and pouring. Carboys are leakproof on standard test conditions. Graduated to identify the volume dispensed. Stopcocks are tied to the handle and require hand assembly for use. No tools are necessary.

HDPE carboys
These high-density polyethylene (HDPE), disposable carboys are ideal for storing and transfer of sterile fluids and pharmaceutical/biotech reagents.
Good for cold storage (down to −80 °C), working temperature of −80 to +120 °C
High impact strength, even at negative temperatures
Widely used for long storage due to high chemical resistance property
Widely used in food and milk industries

PP carboys
Ideal for sterile water storage, polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Excellent chemical resistance
Working temperature: 0 to +135 °C
Autoclavable

Note: Polypropylene carboys are autoclavable. HDPE carboys are not autoclavable. Always remove stopcock and cap from the carboy before autoclaving.

Certifications: Resin meets US FDA 21 CFR, USP Class VI
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR