Carboys, narrow neck, round, with screw caps, VWR®

Supplier: VWR, part of Avantor
VWR®
216-1737EA 56.4 EUR
216-1737 VWRU10755-114 216-1908 VWRU10755-110 216-1729 216-1704 216-2180 216-1738 216-1736 216-1918 216-1703 216-1706 216-1702 216-1701
Carboys, narrow neck, round, with screw caps, VWR®
Bottles Carboys
LDPE, HDPE or PP, translucent with purple screw caps. Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.

LDPE carboys
Ideal for high purity applications as they have high resistance to chemicals (widely used in ophthalmic product range, like eye droppers and nasal sprays).
Ideal resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impact (will not break even if it falls accidentally, wheras PP carboys may lead to cracking at negative temperatures)
LDPE products will bounce back to its original shape, as it is a highly flexible material
Widely used in pharma industries

HDPE carboys
High density polyethylene (HDPE), disposable carboys are ideal for storing and transfer of fluids and pharmaceutical/biotech reagents.
Good for cold storage (down to −80 °C), working temperature of −80 to +120 °C
High impact strength even at negative temperatures
Widely used for long storage periods due to high chemical resistance
Widely used in food and milk industries

PP carboys
Ideal for sterile water storage, polypropylene (PP) carboys can be autoclaved before refilling to stop recurring bacterial growth.
Excellent chemical resistance
Working temperature: 0 to +135 °C
Autoclavable

Note: Polypropylene carboys are autoclavable. LDPE and HDPE carboys are not autoclavable. Always remove cap from carboy before autoclaving.

Certifications: Resin meets US FDA 21 CFR, U SP Class VI.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR