VWR® Sample Vials, with Snap-Caps
Supplier: VWR
VWR®
216-1849EA 170 EUR
216-1849 216-1848 216-1847 216-1851 216-1846 216-1845 216-1844 216-1843
VWR® Sample Vials, with Snap-Caps
Vials
Soda-lime glass, amber.
  • Ideal for storing medication as liquid, powder or capsules
  • Used for light-sensitive liquids
  • White snap-caps are impervious to ultraviolet light
  • Not suitable for heat applications
Delivery information: Delivered in practical safe trays. PE snap-caps are included.