Jerricans, Nalgene®
Supplier: Thermo Fisher Scientific
Nalgene®
216-8693EA 123 EUR
216-8693 216-8691 216-8692
Jerricans, Nalgene®
Bottles Jerricans
HDPE, translucent with white PP screw cap.
  • Good chemical resistance
  • Temperature resistant from −100 to +120 °C
  • Stable sturdy design with integral graduations in gallons and litres, leakproof closure
Good stability through a large base and low centre of gravity. The spout is long enough for accurate pouring of liquids. With tethered sealing cap that cannot be lost. Recessed base offers additional support during pouring.
  • Good chemical resistance
  • Temperature resistant from −100 to +120 °C
  • Stable sturdy design with integral graduations in gallons and litres, leakproof closure
Good stability through a large base and low centre of gravity. The spout is long enough for accurate pouring of liquids. With tethered sealing cap that cannot be lost. Recessed base offers additional support during pouring.