Spoon spatulas, broad shape
Supplier: Nickel-Electro
231-0156EA 17.8 EUR
231-0156 231-0076 231-0077 231-0075
Spoon spatulas, broad shape
Spatulas
Pure nickel (99,6%).
  • One end is flat and spatula-shaped, the other end is spoon-shaped
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean