Double spatulas, Chattaway type, stainless steel
Supplier: Nickel-Electro
231-0344EA 13.8 EUR
231-0344 NICK2305 231-0078 231-0342 231-0343 231-0340 231-0362 NICK2301 231-0363 231-0341 231-0360 231-0361
Double spatulas, Chattaway type, stainless steel
Spatulas
Multipurpose spatula made of stainless steel for a wide range of uses. One end is flat and spatula-shaped, the other end is rounded and turned up at 45°. Available in light or medium weight design.
  • Very good chemical resistance
  • Very good thermal resistance
  • Quick and easy to clean