Scoops
Supplier: Nickel-Electro
NICK9030EA 57.1 EUR
NICK9030 231-0351 231-0352 231-0350
Scoops
Scoops
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
  • Excellent thermal resistance, autoclavable
  • Non magnetic, bright finish
  • Highly polished surface makes cleaning easier