GC septa, Thermogreen® LB-2
Supplier: Merck
23159.EA 113 EUR
23159. 28021-U 23157. SUPL20654 23156. SUPL20666 20652. 20651. SIAL23154
GC septa, Thermogreen® LB-2
Septa
Ready to use GC septa for inlet temperature of up to 100 to 350 °C.
  • Low bleeding
  • Easy needle penetration
  • High puncture tolerance and ideal for autosamplers