Dropping funnels, cylindrical, without pressure balance
Supplier: Lenz
271-5115EA 86.3 EUR
271-5115 271-5114 271-5103 271-5116 201-0896 271-5113 271-5102 271-5112
Dropping funnels, cylindrical, without pressure balance
Funnels Dropping Funnels
DURAN®, borosilicate glass, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket. With divisions. Stopcock with thread guard.