VWR®, Sampling lance
Supplier: VWR
VWR® MIPAW
300-0163EA 169 EUR
300-0163
VWR®, Sampling lance
Samplers Sample Dippers
Needle sampler suitable for collecting powdered milk, flour, sand and cement in dairy, food, feed and chemical industries.
  • AISI 316L electropolished stainless steel
  • Aluminium handle
  • Sterilisable
Method of use: The sampler is placed into the product to be sampled and then transferred to a container.