Disposable samplers, ViscoDispo
Supplier: Bürkle
ViscoDispo
300-0207EA 147.7 EUR
300-0207 300-0208 300-0209 300-0206
Disposable samplers, ViscoDispo
Samplers Disposable Samplers
This ViscoDispo sampler is for highly viscous media up to 100 000 cP. The ergonomically shaped handle and stable drawbar make the sample easier to draw.
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated