Disposable samplers, DispoPicker
Supplier: Bürkle
DispoPicker
300-0262EA 124.6 EUR
300-0262 300-0260 300-0259 300-0261
Disposable samplers, DispoPicker
Samplers Disposable Samplers
This disposable DispoPicker is used in the sampling of powders and small-grain bulk materials. Because the tip is closed, the sample material cannot trickle back out of the sample lance.
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
Ideally suited for cross-section samples.