Disposable samplers, DispoTube
Supplier: Bürkle
DispoTube
300-0267EA 93.8 EUR
300-0267 300-0268 300-0270 300-0269
Disposable samplers, DispoTube
Samplers Disposable Samplers
This DispoTube sampler can be used for taking samples of liquids. Its ergonomic handle makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves.
  • All-layer and target point samples possible
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated