Bottles, wide neck, PET, round or square with screw cap, VWR® Standard Line
Supplier: VWR, part of Avantor
VWR®
331-0167EA 79.3 EUR
VWRI331-0158 331-0159 331-0170 331-0145 331-0167 331-0155 331-0166 331-0143 331-0165 331-0164 331-0163 331-0140 331-0161
Bottles, wide neck, PET, round or square with screw cap, VWR® Standard Line
Bottles Plastic Bottles
PET, clear.
  • 34 mm neck for all bottles
  • Tamper-evident screw caps
  • Manufactured from food grade PET
For basic sample collecting. Non sterile, lightweight, round or square. All caps are HDPE.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904