Water sampling bottles, sterile, VWR®
Supplier: VWR, part of Avantor
331-0256EA 236 EUR
331-0256 331-0273 331-0271 331-0268 331-0272 331-0267 331-0275 331-0274 331-0270 331-0276 331-0277 331-0269
Water sampling bottles, sterile, VWR®
Bottles Water Sampling Bottles
PET, virgin, food grade with PP caps.
  • Different coloured caps for easy indentification of test
  • Tamper-evident strip to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Rectangular bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate
Red cap: For chlorinated water dosed with 20 mg/L sodium thiosulphate
White cap: For chlorinated water dosed with 120 mg/L sodium thiosulphate