Sterile samplers, VWR®
Supplier: VWR Collection
331-0285EA 139 EUR
331-0285 331-0284
Sterile samplers, VWR®
Samplers Sample Dippers
This sampling bottle with removable handle is made of shatter-proof PP.
  • Transparent for immediate visual inspection
  • Red screw cap to ensure leak resistance
  • Hook end to provide a better grip
  • Irradiated
Lot number, expiry date, instruction for use and code number are printed on each bag.
Packaging: Individually wrapped.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904