Water sampling bottles, with tamper evident cap
Supplier: VWR, part of Avantor
VWR®
331-0395EA 127 EUR
331-0395 331-0389 331-0391 331-0383 331-0400 331-0390 331-0392 331-0386 331-0401 331-0394 331-0396 331-0393 331-0384 331-0397 331-0398 331-0399 331-0387 331-0402 331-0388 331-0385
Water sampling bottles, with tamper evident cap
Bottles Water Sampling Bottles
HDPE water sampling collection bottle.
  • Bottle made with HDPE resin
  • Natural bottle colour, with red cap
  • Supplied sterile
  • Sodium thiosulphate in powder form
  • Product labelled with unique barcode and sample collection write-on for traceability
  • Tamper-evident cap options with 33 mm pull type, 45 and 63 mm threaded cap
Wide neck design allows for easy sample collection. Product labelled with dosage and write-on details. These bottles are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken. Square or rectangular bottles allow for more efficient storage.
Certifications: Compliance to US FDA 21 CFR. In compliance Dosage with standards: ISO 5667-3/ISO 19458.