Rotors for cell washer
Supplier: Thermo Fisher Scientific
390-0933EA 2210 EUR
390-0933 390-0934
Rotors for cell washer
Centrifuges Rotors
Meets protocol requirements with the flexibility to run 12 or 24 samples and finish washing in as little as 3 minutes with Thermo Scientific™ cell washer rotors.
  • For washing, tubes swing at a fixed positive angle to sediment cells rapidly at the bottom of the tube
  • For decanting, the rotor holds tubes at a slightly negative angle and the saline is decanted centrifugally
  • Saline distributor mounted directly on top of the rotor to reduce risk of cross-contamination
  • Easy to remove for cleaning convenience
  • Stainless steel rotor body advances system durability