Sterile containers, Falcon®
Supplier: Corning
Falcon®
392-0298EA 353.14 EUR
392-0298 392-0299 392-0396 391-0020 391-0023
Sterile containers, Falcon®
Sample Containers General Purpose Containers
PP, sterile, with or without PE lid.
  • Moulded-in graduations for easy measurement
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904