Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Supplier: Corning
Falcon®
391-1998EA 161.23 EUR
391-1998 391-1997 391-2266 391-2002 391-2003
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Culture Dishes Petri Dishes
Dishes are constructed with crystal-grade polystyrene and a flat bottom surface for distortion-free optics.
  • Flat, distortion free optics
  • Stacking rings allow for easier stacking and handling
  • Durable construction for stable dish manipulation
Small diameter saves media and space. Easily sealed with tape for collecting and transporting specimens. Polystyrene. Sterilized by gamma irradiation. Packed 20 per bag.