Reagent/centrifuge tube with screw cap
Supplier: Greiner bio-one
391-3450EA 368 EUR
391-3450 525-0396 525-0197 525-0384 391-3460 391-3477 525-0399
Reagent/centrifuge tube with screw cap
Tubes Centrifuge Tubes
PP with HDPE screw cap.
  • Excellent thermal, mechanical and chemical stability
  • Blue graduation and white labelling area
  • DNase-free, RNase-free, human DNA-free, pyrogen-free, non-cytotoxic
Universal tubes for a wide range of applications. With or without skirt. Sterile. Autoclavable without a cap.
Packaging: Available in bulk in bags, or packed in boxes.