Test tubes
Supplier: Greiner bio-one
212-3150EA 116 EUR
212-3150 GREI115201_2000 212-3140 212-0363 212-3186 212-3165 212-3110 212-0361 391-3469 212-3146
Test tubes
Tubes Test Tubes
PS, clear or PP, transparent.
  • PP: good temperature resistance, good chemical resistance
  • PS: highly transparent, well suited to visual measurements
  • Matching push-in stoppers are optionally available
  • Can also be used as centrifuge tubes
Universal tubes for a wide range of laboratory and production applications.