PCR tubes for MicroRack tube system
Supplier: Greiner bio-one
391-3724EA 63.5 EUR
391-3724 391-3725 391-3722 391-3723 391-3721
PCR tubes for MicroRack tube system
Tubes Test Tubes
PP tubes designed to work with the MicroRack tube storage system.
  • Polypropylene tubes have particularly good thermal, chemical and mechanical stability
  • Ideally suited for the storage of chemical and biological samples