Sterile containers, Falcon®
Supplier: Corning Life Sciences
Falcon®
391-0023EA 101.45 EUR
391-0023 392-0298 392-0299 392-0396 391-0020
Sterile containers, Falcon®
Sample Containers General Purpose Containers
PP, sterile, with or without PE lid
  • Moulded-in graduations for easy measurement
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904