Cell culture roller bottles
Supplier: Corning
Corning®
734-1729EA 260.06 EUR
734-1729 392-0307 734-4189 734-1884 734-0095 734-1720 734-1898 734-4191 734-1870 734-0096 392-0286 734-1714 CORN430849_COS CORN430195_COS 734-1730 734-4190 734-1702
Cell culture roller bottles
Bottles Roller Bottles
Roller bottles available with smooth or ribbed surface design for expanded culture area. All bottles are treated for optimal cell attachment. The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields. All bottles are graduated, unless otherwise stated.
  • One piece seamless contruction
  • All bottles have printed lot numbers to aid product traceability
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
  • Ribbed design provides twice the surface area with the same exterior dimensions
Note: Corning recommends 0,2 to 0,3 ml of medium per cm² of growth area, and setting roller rack speeds to provide 0,5 to 1,0 rpm.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904