GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
GelRed® & GelGreen®
41005-1EA 2810 EUR
41005-1 41003-T 41004. 41002-1 41003-1 730-1535 41002. 41003. 41001.
GelRed® & GelGreen® Nucleic acid gel stains
Stains Electrophoresis Stains
GelRed and GelGreen are highly sensitive DNA/RNA gel stains made safe by preventing penetration into cells.
  • Highly sensitive either as precast gel stains or post gel stains
  • Can be microwaved and stored at room temperature
  • GelRed™ has been shown to be less mutagenic than ethidium bromide
  • No destaining necessary
GelRed is compatible with any standard 312 nm transilluminator.
GelGreen can be used with a 254 nm UV transilluminator or gel reader with visible light excitation.