J.T.Baker®, Plate 96
Supplier: Avantor
J.T.Baker®
43001-0119EA 99.53 EUR
43001-0119 43001-0117 43001-0118 43001-0600
J.T.Baker®, Plate 96
Microplates Deep Well Microplates
J.T.Baker Plate 96 round well plates made from PS are ideal for ELISA applications.
  • Alphanumeric coding assists with identification of samples
  • Plate shape significantly reduces sample carry-over
  • Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines