J.T.Baker®, Plate 384
Supplier: Avantor
J.T.Baker®
43004-0200EA 176.33 EUR
43004-0200 43004-0120
J.T.Baker®, Plate 384
Microplates Deep Well Microplates
Available in PP for automated screening and storage systems.
  • Alphanumeric code to assist sample identification
  • Square wells for increased volume
  • V-bottom for high sample recovery
  • High frame (3 mm) for stable and slip-free stacking
J.T.Baker Plate 384 is ideal for small sample volumes.