VWR®, Magnetic Wing Stirrers, Micro
Supplier: VWR
VWR®
442-0496EA 40.9 EUR
442-0496 442-0497 442-0493 442-0494 442-0495
VWR®, Magnetic Wing Stirrers, Micro
Stirrers Magnetic Stirring Bars
PTFE covered, Alnico V magnetic core. Smooth surface. For stirring in test tubes, centrifuge tubes and micro vials. The wings can also be trimmed to size if needed.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)