Magnetic hotplate stirrers, VMS Advanced series
Supplier: VWR, part of Avantor
442-0669EA 730 EUR
442-0669 442-0665 442-0666 442-0667 442-0668 442-0661 442-0662 442-0663 442-0664
Magnetic hotplate stirrers, VMS Advanced series
Stirrers Magnetic Hotplate Stirrers
Compact stirrers with a ceramic glass plate provide excellent chemical resistance. Units feature digital error code display, a 'hot' top indicator to warn users that surface is hot, a fixed safety circuit that shuts off the heater if temperature rises above 550 °C, and three operating modes to choose from (standard, safe and adjustment protection). Stirrers have a speed display scale and optional capability to connect directly to a PT1000 temperature sensor; when using this functionality, control accuracy in the medium is ±0,5 °C.
  • Exact temperature setting via digital potentiometer
  • Simultaneous digital display of target and actual temperatures via LCD
  • Display of actual temperature value in medium with 0,1 °C resolution when using PT1000 temperature sensor
  • Stirrer can be controlled using two microcontrollers
Certifications: Protection class according to DIN EN 60529: IP 21
Delivery information: Supplied with PT1000 probe. A wide range of optional accessories are available.