Low profile laboratory shaker with digital display and a large platform. Continuous operation is guaranteed even under extreme loads. Suitable for vessels which need to be laid flat, for example separating funnels. A variety of different vessels can be accommodated by choosing from the range of optional holding attachments.
- Infinitely variable speed control
- Continuous operation or timer operation from 0 to 56 min
- Electronic speed control
- Can be used at 5 to 50 °C (80% relative humidity), in an incubator or cold room
Certifications: IP protection class according to DIN EN 60529: IP 21
Accessories information: Shaker HS 501 digital accommodates a maximum of 50×25, 48×50, 25×100, 16×250 or 12×500 ml flasks.
Ordering information: Supplied without attachments, required accessories must be ordered separately.