Magnetic hotplate stirrers, ARE/AREX 6-series
Supplier: VELP Scientifica
VELPSA20500460EA 609 EUR
VELPSA20500460 442-1259 442-1317 442-1256 442-1258 VELPSD20500460
Magnetic hotplate stirrers, ARE/AREX 6-series
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrers with resistant die-cast housing that protects the internal parts. The control panel is protected from spills thanks to the run-off groove. A white LED with black background display and a set of icons provide precise settings and clear readout of the working conditions.
  • Brushless motor, maximum speed of 1700 min⁻¹
  • Powerful Alnico magnet with superior magnetic coupling
  • SpeedServo™ torque compensation technology maintains constant speed as viscosity changes
  • Superior thermoregulation both with digital and analogue control
The ARE-6 has an aluminium alloy top plate, analogue speed and temperature controls.
The AREX-6 has a fast heating CerAlTop™ plate which protects from chemicals, scratches and enhances the life time of the unit. Upgradable to digital with the connection of VTF or VTF EVO external thermoregulators.
The AREX-6 Digital has an integrated timer, from 1 minute up to 99 hours or unsupervised operation. Two independent safety circuits automatically switch off the heating in event of over-temperature. State-of-the-art thermoregulation: Stable and fast with minimal overshoot. The powerful PID thermoregulation software allows the operator to set the probe parameters in two different modes: Fine (optimised low temperature overshoot) or, fast (with higher overshoot). A Service Life Meter records the working time of both the motor and the heating element. The operator can set the auto-reverse option for enhanced kinetics at a preferred time interval. The probe icon is illuminated when the PT100 or VTF/VTF EVO is connected. Adjustable temperature limit, if the probe is not correctly immersed in the medium the instrument stops heating up and displays a probe alarm.
The AREX-6 Digital PRO has the same features as the AREX-6 Digital, this unit also includes a connection for external digital thermoregulators.
Please see separate product page for full details of AREX-6 Connect PRO models.
Certifications: IP protection class according to DIN EN 60529: IP 42, CNemkoUS certified.
Ordering information: ARE-6 is supplied without accessories. AREX-6 has choice of stirrer only or with VTF digital thermoregulator or VTF EVO. The AREX-6 Digital is supplied with a PT100 probe. The AREX-6 Digital PRO is supplied with a VTF or VTF EVO. Optional silicone cover protects units against spills. A wide range of accessories are available, please order these separately.