Reciprocating shakers, lab scale, SSL2
Supplier: Cole Parmer
stuart®
444-0114EA 2950 EUR
444-0114
Reciprocating shakers, lab scale, SSL2
Shakers Reciprocating Shakers
Laboratory scale platform shakers, which are ideal for extractions. Units have a powerful yet quiet shaking mechanism and have been designed for problem-free continuous use. The cradle type platform has four rubber cushioned horizontal securing bars, which can be easily adjusted horizontally or vertically, to hold different flasks, bottles and beakers. Shakers can accommodate Erlenmeyer flasks or bottles, up to 12×250, 9×500, 6×1000 or 2×2000 ml. Speed is fully variable, and is set digitally ensuring consistency, the speed accuracy is maintained even over long runs.
  • Fully adjustable cradle system holds different sizes and types of vessel
  • Digital speed selection up to 250 rpm with soft start
  • Can be used at 4 to 40 °C (80% relative humidity), in a CO₂ incubator or environmental chamber
  • Digital timer which can be set can be set in hours, minutes, seconds or for continuous operation, with acoustic signal
  • BioCote® antimicrobial protection
IP Protection Class according to DIN EN 60529: IP 31