Magnetic hotplate stirrers, Corning®
Supplier: Corning
Corning®
444-1017EA 0 EUR
444-1017 444-1005 444-1002 CORN6795-220 444-1014 444-1011 444-1001 444-1008 CORN6795-420D
Magnetic hotplate stirrers, Corning®
Stirrers Magnetic Hotplate Stirrers
These hot plate stirrers have a durable glass-ceramic Pyroceram® top and offer efficient performance and reliability in a low profile, space-saving design.
  • Exclusive closed-loop stirring control monitors and regulates the stirring speed for aqueous, viscous, or semi-solid solutions
  • Microprocessor maintains consistent and repeatable temperature settings
  • For digital displays, LED temperature display is adjustable in increments of 5 °C and flashes until set temperature is reached
  • Built-in holder accommodates optional 102 mm stainless steel support rods
  • Stronger, larger magnet improves magnetic stirring bar coupling combined with automatic ramp up speed adjustment which minimises decoupling
  • Optional external temperature controller truly controls temperature inside the vessel
Available with a choice of two top plate sizes, these feature an LED “hot top” icon that indicates when the plate temperature is over 60 °C, even when heat control is off. Separate temperature sensors will provide power cutoff if the unit overheats. LED temperature display is adjustable in 5 °C increments, and blinks until set temperature is reached.
Certifications: UL/cUL approved.
Delivery information: All units include manufacturer's two-year warranty. Stirring hot plate kit contains unit plus temperature controller, support rod, and stir bar retriever. The optional external temperature controller ensures precision temperature accuracy of the liquid inside the vessel. All accessories are otherwise sold separately.