Reciprocating shakers, Promax
Supplier: Heidolph
444-1478EA 1474 EUR
444-1478 444-1420 444-1255 444-1256 444-1473 444-1320
Reciprocating shakers, Promax
Shakers Reciprocating Shakers
Promax shakers have a one dimensional back and forth motion for vigorous mixing of flasks and separating funnels in chemical applications, using analogue dial controls for reciprocating frequency. The encapsulated housing ensures virtually no heat transfer from motor to platform and a sealed drive is isolated from spills.
  • Speed continuously adjustable
  • Modular system; combine Promax 1020 with Incubator 1000 with choice of a low or high incubator hood
  • Process timer with audible signal allows for unattended operation
  • Promax 1020; digital timer up to 999 min, Promax 2020; analogue timer up to 120 min
  • Can be used at 0 to 50 °C (80% relative humidity), in an incubator or cold room
Certifications: IP protection class according to DIN EN 60529: IP 40 (Promax 1020); IP 20 (Promax 2020)
Delivery information: Supplied with non-slip rubber mat. Large range of attachments and clamps for up to four separating funnels, Erlenmeyer flasks and tension rollers are available separately. Promax 1020 accessories are category D, Promax 2020 accessories are category E, see entry for 'Accessories for Heidolph shakers' towards the end of the orbital shakers section. Please contact your local Avantor office for further details.