Digital shaker with microprocessor control which is ideal for a range mixing applications, e.g. solubility studies and extraction procedures. Variable speed control provides consistent, uniform shaking action. The shaking system continuously monitors shaking speed and will maintain the set point even under changing loads.
- Two independent LED displays show speed and time
- Integral timer, 1 s to 160 h, shows elapsed time or counts down, automatic switch off and audible alarm when program is complete
- Can be used at −10 to +60 °C (maximum 80% relative humidity, non condensing), in an incubator, CO₂ incubator or cold room
Displayed speed is accurate to 1% of set speed (when speed is above 100 min⁻¹). The ramping feature slowly increases speed to the set point for added safety. Built-in load sensor can detect an unbalanced condition and will automatically reduce the min⁻¹ to a safe speed to prevent spills and protect samples. Speed calibration mode allows user to recalibrate the speed display and RS232 serial port provides two-way communication for data logging and unit control.
Delivery information: Supplied with non-slip rubber mat, but without attachments, accessories must be ordered separately.
Caution: * Platforms are suitable for optional 2-tier braces, (444-2965) which allow stacking with 254 mm clearance between platforms (recommended for selected platform sizes)
** Platform stacking is not recommended for 3750 or 5000 models