Burner, Teclu
Supplier: Usbeck
451-1343EA 47.2 EUR
451-1343 USBE1301 451-0088 451-0087 451-1366 451-1341 USBE1200 451-0086 USBE1201
Burner, Teclu
Heaters Gas Burners
Nickel plated brass base, in powder coated grey.
  • With air regulation valve and throttle screw
  • Without pilot light
Height: 165 mm