Bunsen burners
Supplier: Usbeck
451-0082EA 48.1 EUR
451-0082 451-0081 451-0168
Bunsen burners
Heaters Gas Burners
Nickel plated brass with cast iron foot.
  • With air regulation and pilot flame
  • Lever stop-cock
Height: 150 mm.
Ordering information: Supplied without the black rubber safety base that is shown in the image. This optional protective base (451-1342) must be ordered separately.