Bunsen burners
Supplier: Usbeck
451-0078EA 29.5 EUR
451-0078 451-0083 451-1368 451-0075 451-0079 451-0077 451-0076 451-0080 451-1367
Bunsen burners
Heaters Gas Burners
Nickel plated brass with cast iron foot.
  • With air regulation ring
Height: 145 mm (451-0079, 451-0080: 150 mm, 451-0083: 160 mm)
Head Ø: 17 mm (14 mm for propane models)