Baths, thermostatically controlled, analogue, Clifton NE4-T series
Supplier: Nickel-Electro
Clifton
462-0107EA 1300 EUR
462-0107 462-0108 462-0109 462-0110
Baths, thermostatically controlled, analogue, Clifton NE4-T series
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
These thermostatically controlled analogue stirred baths have a corrosion resistant stainless steel tank and durable outer case. Immersed components are corrosion resistant.
  • Thermostatic control, easy to use dial setting, ambient 5 to 99 °C
  • Powerful stirring within the tank for good water circulation and temperature stability
  • Controls with an illuminated on/off switch and 'heater on' indicator
  • User resettable safety cut-out float switch for low liquid level protection
  • Anti-bacterial paint finish that actively inhibits bacterial growth
*RT = Ambient