Circulators, NE4 series, Clifton
Supplier: Nickel-Electro
Clifton
462-0113EA 1950 EUR
462-0113 462-0047 462-0114 462-0049 462-0039 462-0044 462-0112
Circulators, NE4 series, Clifton
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
These circulating water baths are available in two ranges for circulating just water up to 99 °C or oil up to 130 °C. Operating temperatures below ambient can be achieved using an optional refrigerated dip cooler. Units have touch pad controls with easy wipe clean fronts which are chemical resistant and splash proof.
  • Powerful circulation pump, provides a constant flow
  • Convenient ‘push-fit and lock' fittings on both inlet and outlet ports for secure external water circulation
  • Control options, single point temperature control or process control with two temperatures and a time setting
  • Automatic setting alarms for protecting bath contents, audible and visual indication
  • Anti-bacterial paint finish that actively inhibits bacterial growth
Maximum pump capacity:
10 L/min, 147 mbar
*RT = Ambient
Ordering information: Supplied with a stainless steel mounting bridge and removable stainless steel shelf, but without racks or lids (lids are recommended when working above 60 °C or below ambient). Optional dip cooler is required for operating at below ambient temperature. Please order required accessories separately.