Desiccator lids, DURAN®
Supplier: DWK Life Sciences
467-1375EA 105.09 EUR
467-1375 467-1390 467-1280 467-1290 467-1180 467-1260 467-1380 467-1160 467-1185 467-1385 467-1275 467-1285 467-1175 467-1190
Desiccator lids, DURAN®
Lids Desiccator Lids
DURAN®, borosilicate glass 3.3, clear plain lids for desiccators. Lids have either knob or NS-connection for stop cork (Novus) or thread connection for stop cork (Mobilex).
Certifications: According to DIN ISO 13130