Autoclaves, LABOKLAV
Supplier: SHP
481-0459EA 27460 EUR
481-0459 481-0415 481-0416 481-0417 481-0418 481-0419 481-0495 481-0451 481-0452 481-0453 481-0454 481-0410 481-0411 481-0455 481-0456 481-0412 481-0457 481-0413 481-0414 481-0458 481-0460 481-0461 481-0426 481-0427 481-0428 481-0429 481-0629 481-0462 481-0463 481-0420 481-0464 481-0465 481-0421 481-0422 481-0466 481-0423 481-0467 481-0468 481-0424 481-0425 481-0469 481-0470 481-0471 481-0472 481-0437 481-0438 481-0439 481-0473 481-0430 481-0474 481-0475 481-0431 481-0476 481-0432 481-0477 481-0433 481-0434 481-0478 481-0479 481-0435 481-0436 481-0480 481-0481 481-0482 481-0483 481-0448 481-0404 481-0449 481-0405 481-0406 481-0407 481-0408 481-0409 481-0440 481-0484 481-0441 481-0485 481-0442 481-0486 481-0487 481-0443 481-0444 481-0488 481-0445 481-0489 481-0401 481-0446 481-0402 481-0447 481-0403 481-0490 481-0491 481-0492 481-0493 481-0450 481-0494
Autoclaves, LABOKLAV
Sterilisers Autoclaves (Steam Sterilisers)
A completely new concept allows the installation of the unit as either front or top loader. Deciding on horizontal or vertical is no longer the question with SHP autoclaves. No matter if installed horizontally or vertically, the units remain the same. Even at a later stage the unit can be changed from vertical to horizontal installation or vice versa without the need for big technical changes (contact Avantor for details). When set up as horizontal units they are even stackable. Stainless steel 316L chamber and housing.
A completely new concept allows the installation of the unit as either front or top loader. Deciding on horizontal or vertical is no longer the question with SHP autoclaves. No matter if installed horizontally or vertically, the units remain the same. Even at a later stage the unit can be changed from vertical to horizontal installation or vice versa without the need for big technical changes (contact VWR for details). When set up as horizontal units they are even stackable.
  • Automatic feed water pump
  • Fully automatic closing system and thermal locking device to protect the operator
  • Reference sensor as standard which measures the temperature inside the bottles
  • Microprocessor control with 22 programs that can be adjusted by the operator to optimise the process (code protected)
  • RS485 interface
  • Three colours available: Light grey/blue, light grey/red, light grey/green (please indicate the desired colour when ordering)
  • All models are equipped with a cooling system to ensure steam is cooled prior to evacuation, thus ensuring drain protection
The newly developed integrated steam generator separates the heating elements from the water; the result is a much longer life span of heating elements. Low in energy consumption due to optimised size of steam generator in relation to chamber volume.
Units with cooling or vacuum options are supplied with full double jacket made from the same material as the chamber. During the cooling cycle, deionised water is circulated around the double jacket to ensure a fast and reliable cooling of sensitive media. There is no consumption of cooling deionised water, it returns to the storage tank which has an integral heat exchanger. During the drying cycle the jacket is filled with steam to support the drying of the vacuum unit.
  • Automatic feed water pump
  • Fully automatic closing system and thermal locking device to protect the operator
  • Reference sensor as standard which measures the temperature inside the bottles
  • Microprocessor control with 22 programs that can be adjusted by the operator to optimise the process (code protected)
  • RS485 interface
  • Three colours available: Light grey/blue, light grey/red, light grey/green (please indicate the desired colour when ordering)
  • All models are equipped with a cooling system to ensure steam is cooled prior to evacuation, thus ensuring drain protection
Autoclaves for use in the laboratory, in full compliance with: DIN 58950 (pharmaceutical applications), DIN 58951 (laboratory applications), pressure Device Directive 97/23/EU, IEC 61010 - 2 - 43 (thermal locking device).
Certifications: Autoclaves for use in the laboratory, in full compliance with: DIN 58950 (pharmaceutical applications), DIN 58951 (laboratory applications), pressure Device Directive 97/23/EU, IEC 61010 - 2-43 (thermal locking device).